Order Entry
Norway
ContactUsLinkComponent
 
Irritant
(R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane ≥94.0% (by GC)
Catalog # TCIAH1188-1G
Supplier:  TCI
CAS Number:  
Irritant
(R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane ≥94.0% (by GC)
Catalog # TCIAH1188-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 94.0 %

Specification Test Results

About this item

 :
 :146.19 g/mol
Boiling Pt:110°C
 :GHS07
CAS Number:70005-89-9
Formula:C₇H₁₄O₃
MDL Number:MFCD02682966