
(4-Hydroxyphenyl)diphenyl methanol ≥98.0% (by HPLC)
Catalog # TCIAH1059-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:160.0 to 164.0 deg-C
Specification Test Results
About this item
:
:276.34 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:15658-11-4
Formula:C₁₉H₁₆O₂
MDL Number:MFCD00973626