m-PEG7-alcohol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4437-01-8
:
Formula:C₁₅H₃₂O₈
MDL Number:MFCD06201006
:340.41 g/mol
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %