Order Entry
Norway
ContactUsLinkComponent
 
N-Hydroxy-1,8-naphthalimide ≥97.0% (by HPLC, titration analysis)
 :  TCI
 
N-Hydroxy-1,8-naphthalimide ≥97.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %