Order Entry
Norway
ContactUsLinkComponent
Hexahydroxybenzene ≥98.0% (by titrimetric analysis)
Irritant
Hexahydroxybenzene ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0643-1G
Supplier:  TCI
CAS Number:  
Irritant
Hexahydroxybenzene ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0643-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (Cerium redox Method):
    min. 98.0 %

Specification Test Results

About this item

 :
 :174.11 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:608-80-0
Formula:C₆H₆O₆
MDL Number:MFCD00013966