Sodium-4-hydroxy-3-methoxybenzoate ≥98.0% (by GC, titration analysis)
: TCI
:28508-48-7
:
:C₈H₈O₄
:MFCD00016534
:190.13 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency