
Gabapentin ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:60142-96-3
:
Formula:C₉H₁₇NO₂
MDL Number:MFCD00865286
:171.24 g/mol
Merck Index:14,04319
Melting Pt:166°C
EINECS:262-076-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together