
(Fmoc-amino)-PEG12-C2-Carboxylic acid
Catalog # TCIAF1238-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:839.97 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1952360-91-6
Formula:C₄₂H₆₅NO₁₆
MDL Number:MFCD08064302