Order Entry
Norway
ContactUsLinkComponent
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    186.0 to 190.0 deg-C

Specification Test Results