Order Entry
Norway
ContactUsLinkComponent
 
N-Fmoc-L-homoarginine ≥97.0% (by HPLC, titration analysis)
 :  TCI
 
N-Fmoc-L-homoarginine ≥97.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %