Fludarabine monophosphate ≥98.0% (by HPLC, titration analysis)
: TCI
:75607-67-9
:
:C₁₀H₁₃FN₅O₇P
:MFCD00866418
:14,04126
:203°C
:365.21 g/mol
:GHS07
:GHS08
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+11.0 to +13.0 deg (C=0.5, H2O)