Order Entry
Norway
ContactUsLinkComponent
Fenbendazole ≥98.0% (by HPLC, total nitrogen)
Fenbendazole ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAF0812-25G
Supplier:  TCI
CAS Number:  
Fenbendazole ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAF0812-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

About this item

 :
 :299.35 g/mol
Merck Index:14,03960
Melting Pt:233°C
EINECS:256-145-7
CAS Number:43210-67-9
Formula:C₁₅H₁₃N₃O₂S
MDL Number:MFCD00144301