1-Ethylcyclopentyl acrylate ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:326925-69-3
:
Formula:C₁₀H₁₆O₂
MDL Number:MFCD31716062
:168.24 g/mol
:Stabilised with 4-methoxyphenol (MEHQ)
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %