Order Entry
Norway
ContactUsLinkComponent
 
 
 
(5-Ethyl-1,3-dioxan-5-yl)methyl acrylate ≥98.0% (by GC) stabilised
 :  TCI
 
undefined
 
 
(5-Ethyl-1,3-dioxan-5-yl)methyl acrylate ≥98.0% (by GC) stabilised
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Acid Value:
    max. 2.0

 

 

Frequently Bought Together