
(2-Ethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:70289-12-2
:
Formula:H₃CCH₂O(C₆H₄)CH₂COOH
MDL Number:MFCD00016824
:180.2 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:103.0 to 107.0 deg-C