2-Ethynylbenzothiazole ≥97.0% (by GC)
Catalog # TCIAE1091-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:39.0 to 43.0 deg-C
Specification Test Results
About this item
:
:159.21 g/mol
Storage Temperature:Freezer
CAS Number:40176-80-5
Formula:C₉H₅NS
MDL Number:MFCD08703599