Order Entry
Norway
ContactUsLinkComponent
 
 
4-Ethylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
  TCIAE0966-5G
 :  TCI
 :  
 
4-Ethylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
  TCIAE0966-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Neutralization titration):
    min. 98.0 %

 

 

 :
 :156.23 g/mol
 :GHS07
 :91328-77-7
 :C₉H₁₆O₂
 :MFCD01568857