
Ethyl-1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:100501-62-0
:
Formula:C₁₄H₁₂F₃NO₃
MDL Number:MFCD00276022
:299.25 g/mol
EINECS:405-880-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:205.0 to 209.0 deg-C