
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2199-44-2
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00010645
Boiling Pt:272°C
:167.21 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:122.0 to 126.0 deg-C