N-Ethoxycarbonyl-5-nitro-o-toluidine ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:16648-52-5
:
Formula:C₁₀H₁₂N₂O₄
MDL Number:MFCD00059827
:224.22 g/mol
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:132.0 to 136.0 deg-C
- Solubility in hot Methanol:almost transparency