Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethylene glycol mono-m-tolyl ether
Ethylene glycol mono-m-tolyl ether ≥98.0% (by GC)
Ethylene glycol mono-m-tolyl ether ≥98.0% (by GC)
:
TCI
Ethylene glycol mono-m-tolyl ether ≥98.0% (by GC)
:
TCI
:
13605-19-1
:
:
C₉H₁₂O₂
:
MFCD00014035
:
152.19 g/mol
:
146°C
:
237-091-3
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 98.0 %