4-(4-Ethoxyphenoxycarbonyl)phenyl ethyl carbonate ≥98.0% (by GC)
TCIAE0257-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:105.0 to 108.0 deg-C
:
:330.34 g/mol
:33926-25-9
:C₁₈H₁₈O₆
:MFCD00059379