Order Entry
Norway
ContactUsLinkComponent
N-Ethyl-2-naphthylamine hydrobromide ≥98.0% (by GC, titration analysis)
N-Ethyl-2-naphthylamine hydrobromide ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
N-Ethyl-2-naphthylamine hydrobromide ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Solubility in dilute HCl:
    almost transparency

Specification Test Results