EDTA magnesium disodium salt hydrate ≥99.0% (by titrimetric analysis)
Catalog # TCIAE0094-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (Chelometric Titration):min. 99.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:16.0 to 20.0 %
Specification Test Results
About this item
:
:358.5 g/mol (anhy)
EINECS:238-372-3
CAS Number:14402-88-1
:MFCD00078217
Formula:C₁₀H₁₂MgN₂Na₂O₈·xH₂O
MDL Number:MFCD03613601