
6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1147124-23-9
:
Formula:C₃₂H₄₀Br₂N₂O₂
MDL Number:MFCD28125575
:644.49 g/mol
:GHS07
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:110.0 to 114.0 deg-C