10-[4-[4,6-Di(adamantan-1-yl)-1,3,5-triazin-2-yl]phenyl]-9,9-dimethyl-9,10-dihydroacridine ≥99.0% (by HPLC), purified by sublimation
Catalog # TCIAD5745-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 99.0 area%
Specification Test Results
About this item
:
:632.90 g/mol
CAS Number:2250187-15-4
Formula:C₄₄H₄₈N₄