(E)-6,6'-Dibromo-[3,3'-biindolinylidene]-2,2'-dione ≥96.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1147124-21-7
Formula:C₁₆H₈Br₂N₂O₂
MDL Number:MFCD29917500
:420.055 g/mol
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 96.0 %