Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethoxyphenol
2,4-Dimethoxyphenol ≥98.0% (by GC)
2,4-Dimethoxyphenol ≥98.0% (by GC)
TCIAD5629-1G
:
TCI
:
2,4-Dimethoxyphenol ≥98.0% (by GC)
TCIAD5629-1G
:
TCI
:
Pk:
1 g/mol
Seq:
9220
Appearance:
White to Orange to Green powder to lump
Purity (GC):
min. 98.0 %
:
:
154.1632 g/mol
:
129°C
:
28°C
:
GHS07
:
13330-65-9
:
C₈H₁₀O₃
:
MFCD00060637