Diethyl 2,4-Dimethyl-5-[(triisopropylsilyl)methylene]-1,3-cyclopentadiene-1,3-dicarboxylate cis- and trans-mixture ≥93.0% (by HPLC)
Catalog # TCIAD5585-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (HPLC):min. 93.0 % (total of isomer)
Specification Test Results
About this item
:
:406.64 g/mol
Storage Temperature:Refrigerator
CAS Number:1417407-35-2
Formula:C₂₃H₃₈O₄Si
MDL Number:MFCD27979856