
2,4-Dichloro-3-nitropyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5975-12-2
:
Formula:C₅H₂Cl₂N₂O₂
MDL Number:MFCD09261088
:192.9876 g/mol
Boiling Pt:78°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C