
2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:461642-78-4
:
Formula:C₁₇H₃₂O₂S₃
MDL Number:MFCD10698871
:364.62 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C