4',5'-Dimethoxy-2'-nitroacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4101-32-0
:
Formula:C₁₀H₁₁NO₅
MDL Number:MFCD00130048
:225.20 g/mol
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:131.0 to 135.0 deg-C
Specification Test Results
Frequently Bought Together