Dansylglycine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Sertifikat
Om dette produktet
CAS-nummer:1091-85-6
:
Formel:C₁₄H₁₆N₂O₄S
MDL-nummer:MFCD00037734
:308.35 g/mol
EINECS:214-129-7
Spesifikasjonstestresultater
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:156.0 to 161.0 deg-C
- Human serum albmin (HSA) binding activity:S/N ratio min. 8