Order Entry
Norway
ContactUsLinkComponent
1,3-Dibromo-5-nitrobenzene ≥98.0% (by GC)
1,3-Dibromo-5-nitrobenzene ≥98.0% (by GC)
  TCIAD5383-1G
 :  TCI
 :  
undefined
1,3-Dibromo-5-nitrobenzene ≥98.0% (by GC)
  TCIAD5383-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    103.0 to 107.0 deg-C

 

 

 :
 :280.9 g/mol
 :GHS07
 :6311-60-0
 :C₆H₃Br₂NO₂
 :MFCD00092529