
2,6-Difluoro-4-iodoaniline ≥98.0% (by GC)
TCIAD5214-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:80.0 to 84.0 deg-C
:
:255.0039 g/mol
:GHS07
:141743-49-9
:C₆H₄F₂IN
:MFCD07774189