Order Entry
Norway
ContactUsLinkComponent
N,N'-Di(9-phenanthrenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
N,N'-Di(9-phenanthrenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
N,N'-Di(9-phenanthrenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results