
1,12-Dodecanediol Dimethacrylate ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:72829-09-5
:
Formula:C₂₀H₃₄O₄
MDL Number:MFCD00080499
:338.49 g/mol
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:276-900-4
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
- Specific gravity (20/20):0.9540 to 0.9580
- Refractive index n20/D:1.4590 to 1.4630