Order Entry
Norway
ContactUsLinkComponent
9,9-Diphenylfluorene-2-boronic acid (contains varying amounts of Anhydride)
Irritant
9,9-Diphenylfluorene-2-boronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
Irritant
9,9-Diphenylfluorene-2-boronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 106.0 %

Specification Test Results

Frequently Bought Together