
2,5-Dichloro-4-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:847664-64-6
:
Formula:C₅H₄BCl₂NO₂
MDL Number:MFCD06798254
:191.81 g/mol
Melting Pt:202°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):96.0 to 111.0 %