1,2-Dimethyl-1,1,2,2-tetraphenyldisilane ≥98.0% (by GC)
TCIAD5031-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:142.0 to 146.0 deg-C
:
:185°C
:394.6557 g/mol
:214-632-1
:1172-76-5
:C₂₆H₂₆Si₂
:MFCD00051536