Order Entry
Norway
ContactUsLinkComponent
 
Di(trimethylolpropane) ≥98.0% (by GC)
 :  TCI
 
Di(trimethylolpropane) ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    109.0 to 113.0 deg-C