9,10-Diphenylanthracene ≥99.0% (by GC), purified by sublimation
Katalog # TCIAD4401-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:250.0 to 254.0 deg-C
Specification Test Results
About this item
:
:330.43 g/mol
Boiling Pt:485°C
EINECS:216-105-1
CAS Number:1499-10-1
Formula:C₂₆H₁₈
MDL Number:MFCD00001253