Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate ≥98.0% (by GC)
Catalog # TCIAD4397-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:326.21 g/mol
CAS Number:54094-19-8
Formula:C₁₂H₁₄F₃O₅P
MDL Number:MFCD09054713