
Diphenylbis(phenylethynyl)silane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18784-61-7
:
Formula:C₂₈H₂₀Si
MDL Number:MFCD00093508
:384.55 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:79.0 to 83.0 deg-C