Order Entry
Norway
ContactUsLinkComponent
 
 
2,5-Dichloro-3-nitropyridine ≥98.0% (by GC)
 :  TCI
 
 
2,5-Dichloro-3-nitropyridine ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    43.0 to 47.0 deg-C