1,2-Dioleoyl-sn-glycero-3-phosphocholine ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAD4250-250MG
Supplier: TCI
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (HPLC) (CAD):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:+4.0 to +6.0 deg (C=0.5,CHCl3:MeOH=4:1)
Specification Test Results
About this item
:
:786.13 g/mol
Storage Temperature:Freezer
EINECS:224-193-8
CAS Number:4235-95-4
Formula:C₄₄H₈₄NO₈P
MDL Number:MFCD00135191