Order Entry
Norway
ContactUsLinkComponent
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Irritant
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Catalog # TCIAD4247-5G
Supplier:  TCI
CAS Number:  
Irritant
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Catalog # TCIAD4247-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Suitability for protein-analysis:
    to pass test

Specification Test Results

About this item

 :
 :GHS07
CAS Number:15178-76-4
Formula:C₁₃H₂₉NO₃S
MDL Number:MFCD00036912