Dimethyl(2-oxoheptyl)phosphonate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:36969-89-8
:
Formula:C₉H₁₉O₄P
MDL Number:MFCD00009514
:222.22 g/mol
Boiling Pt:89°C
EINECS:253-293-4
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %