
2,5-Dichloronicotinic acid ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:59782-85-3
:
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD01861979
:192 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:154.0 to 158.0 deg-C