Order Entry
Norway
ContactUsLinkComponent
Dimethylamine ∼11% ∼2 mol/L in isopropanol (IPA)
Flammable
Corrosive
Irritant
Health
Dimethylamine ∼11% ∼2 mol/L in isopropanol (IPA)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Dimethylamine ∼11% ∼2 mol/L in isopropanol (IPA)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Neutralization titration):
    10.5 to 13.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together